Learn More
Electron Microscopy Sciences EMS Bent Cryo Pucks Tong
SDP
Supplier:  Electron Microscopy Sciences 7116811
Encompass_Preferred
Bent Cryo Tong is used for easy and secured removal of cryo boxes in EMS cryo pucks.
Catalog No. 50-192-9424