Learn More
Electron Microscopy Sciences EMS Cryo Pucks TEM Grid Storage
SDP
Supplier:  Electron Microscopy Sciences 7116802
Encompass_Preferred
Cryo Pucks provide organized storage and transport for round Cryo-EM grids boxes under cryogenic conditions. There are 12 indexed wells in each puck to hold round cryo grid boxes. The pucks have a unique alpha-numeric code for easy identification and sample tracking. The puck depth accomodates pin type lid style cryo grid boxes.
When using cryo grid boxes with flat-style lids, you can store up to 24 cryo grid boxes per puck. Special tweezer slots allow easy and secure removal of cryo grid boxes.
Catalog No. 50-192-9419