Learn More
Andwin Scientific CRYOBABIES RD 33X13 1700/PK
SDP
Supplier:  Andwin Scientific 77560R
Encompass_Preferred
Suitable for laser printing, the 33X13 Cryo-Babies stay-flat labels in Red can be used on 1.5mL tubes. Made of polyolefin and acrylic adhesive, it is chemical and temperature resistant from -196-150 deg.C. It comes in 1700 a pack.
Catalog No. 50-727-674