Learn More
LabScientific Cardboard cell dividers for 100 cells (10x10 format). 12 pack. Fit in Cryovial Boxes. Fits all 2" and 3" cardboard and metal boxes.
Supplier:  LabScientific CBL100
Encompass
Cardboard cell dividers for 100 cells (10x10 format). 12 pack. Exterior dimensions: 4 7/8×4 7/8×1 5/16 inch, Cell opening: 0.45 inch (11.5mm)
Catalog No. NC0639900