Learn More
LabScientific Cardboard cell dividers for 81 cells (9x9 format). 12 pack. Fit in Cryovial Boxes. Fits all 2" and 3" cardboard and metal boxes.
Supplier:  LabScientific CBL81
Encompass
81 Cardboard Cell Divider (9x9), 12/PK Cell Partitions That Fit In Cryovial Boxes. Fits all 2" and 3" cardboard and metal boxes. Exterior dimensions: 4 7/8 × 4 7/8 × 1 5/16 inch (123.4×123.4×33 mm), Cell opening: 0.50 inch. (12.7 mm).
Catalog No. NC9709231