Learn More
Molecular Dimensions CRYOCAPS (PLAIN)
SDP
Supplier:  Molecular Dimensions MD7400
Encompass_Preferred
Protein Crystallography. CryoCap (Plain) 50 (SPINE standard) Holds 18 mm SPINE standard pins precisely and securely. With 18mm pins, sample holder length (cap + pin + loop) is 22mm. Drain hole for liquid nitrogen to facilitate positive docking with the goniometer mount. Compatible with existing goniometer mounts.
Catalog No. NC0361819