Learn More
Molecular Dimensions CRYOCAPS WITH DATA MATRIX 50PK
SDP
Supplier:  Molecular Dimensions MD7401
Encompass_Preferred
Protein Crystallography. CryoCaps with Data Matrix (50)(SPINE standard)With 18mm, sample holder length (cap + pin + loop) is 22mm. DATA MATRIX coded and human readable code for automated sample recording. Drain hole for liquid nitrogen to facilitate positive docking with the goniometer mount.Compatible with existing goniometer mounts.
Catalog No. NC9821745