Learn More
Mitegen Shelved Storage Cane for MiTeGen 2nd Gen Cryo-EM Pucks , Holds 10 Pucks, Compatible with TW34, TW35-6, HC34, VHC35 Dewars
SDP
Supplier:  Mitegen MCEMSTC2
Encompass_Preferred
Shelved Storage cane for the MiTeGen 2nd Generation Cryo-EM Puck SKU M-CEMG2-1. Design to hold up to 10 pucks. The shelved storage cane is compatible with IC Biomedical TW34, TW35-6, HC34 and VHC35 storage dewars. The cane streamlines inventory management and sample retrieval processes by keeping pucks neatly organized and easily accessible. Its robust design contributes to maintaining sample integrity by minimizing exposure during transfers and safeguarding specimens in ultra-low temperature environments. How to use - Once you have filled your grid boxes with samples, place the grid boxes in the cryoem storage puck, then load the cryoem storage pucks into the shelved storage cane. Once you have the pucks in the shelved storage cane, place the cane in the storage dewar. Note, this shelved storage cane is not designed for transport use. If you need cane for transporting your pucks, see SKU M-CEMTPC-1.
Catalog No. NC1863751