Learn More
GA International Cryo-DTermo™ – Chemical-Resistant DYMO-Compatible Cryogenic Labels
Supplier:  GA International EDCC082WH
Encompass_Preferred
DYMO-compatible chemical and ultra-low temperature resistant labels. Ideal for the identification and permanent storage of vials, tubes, and other containers in liquid nitrogen (-196°C/-321°F), laboratory freezers (-80°C/-112°F), and dry ice transportation. Coated with a strong permanent adhesive, these direct thermal labels withstand multiple freeze-thaw cycles and can be immersed in liquid nitrogen directly after application, requiring zero cure time. These cryo labels can also withstand direct exposure to harsh chemicals, solvents, and detergents, such as alcohols, toluene, acetone, and xylene, as well as rubbing with surface sanitizing wipes. Resistant to short-term immersion (up to 3 minutes) in alcohols and xylene.
Catalog No. NC1790009