Learn More
GA International Cryogenic Thermal-Transfer Labels for Frozen Vials & Tubes 0.875" x 0.875" + 0.875" (wrap) / 22.2mm x 22.2mm + 22.2mm (wrap) Rectangle White + Transparent From -196°C to +120°C (-320.8°F to +248°F)
Supplier:  GA International L2FX327NOTC11WH
Encompass_Preferred
Cryogenic labels, with a clear overlap area, designed to be applied directly, and rapidly adhere to frozen plastic, and glass cryovials and tubes, which are difficult to label under deep-freeze conditions (-80°C/-112°F). Designed for long-term preservation in liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers and dry ice transportation, and can withstand temperatures up to +120°C (248°F). Labels can be printed with all common brands of thermal transfer printers to obtain a sharp, smudge-proof printout that is also resistant to most common laboratory chemicals and detergents. They can also be written on with our cryo-markers. On vials and tubes, it is recommended that these CryoSTUCK labels are applied with an overlap of at least 0.25” to ensure proper adherence to the frozen surface.
Catalog No. 50-005-72184