Learn More
Chemglass Life Sciences Chemglass Cryogenic Vials, 3D Barcode, SBS Format, External Thread, Caps On, Sterile (CGN-4765-01)
SDP
Supplier:  Chemglass Life Sciences CGN476501
Encompass_Preferred
Sterile cryogenic vials designed for ultra-low temperature sample storage and automation compatibility. Made from medical-grade cryogenic polypropylene, they feature 3D barcode identification and external-thread caps for secure sealing.
  • Made from medical-grade cryogenic polypropylene that meets USP Class-6 standards.
  • Feature three-code-in-one identification with a 1D side barcode and a 2D bottom barcode.
  • Designed to meet SBS format for automation compatibility.
  • Use external thread and caps-on closure for secure sealing.
  • Sterile by electron beam sterilization; sterility assurance level (SAL) = 10^-6.
  • Recommended for liquid nitrogen storage down to -196 °C.
  • Compatible with automated systems using SBS-format racks; racked status not specified for this exact item.
  • Capacity not specified on the manufacturer's page for this exact item.
Catalog No. 50-273-3354