Learn More
Chemglass Life Sciences Cryogenic Vials, 4.0 mL, 3D Barcode, External Thread, Caps On, Racked, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN476504
Encompass_Preferred
Sterile 4.0 mL cryogenic polypropylene vials supplied racked in SBS format for automated handling. The two-color vial body and integrated 3D barcode provide multi-code traceability for sample tracking. Vials are DNase/RNase free, non-endotoxin, and electron beam sterilized (SAL = 10^-6). Supplied 48 per pack, 480 per case.
  • 4.0 mL capacity.
  • Medical-grade cryogenic polypropylene (PP), meets USP Class-6 standards.
  • External thread with caps on.
  • Integrated 3D barcode with 2D bottom code and 1D side code.
  • Sterile; electron beam sterilized, SAL = 10^-6.
  • DNase/RNase free and non-endotoxin.
  • Racked in SBS format; 48 per pack, 480 per case.
Catalog No. 50-273-3351