Learn More
Chemglass Life Sciences Cryogenic Vial, 4.0 mL, 3D Barcode, External Thread, Caps Separated, Sterile (SBS Format)
SDP
Supplier:  Chemglass Life Sciences CGN476506
Encompass_Preferred
Sterile 4.0 mL cryogenic vials designed for long-term liquid-nitrogen storage and automated sample tracking. Medical-grade cryogenic polypropylene construction and a three-code marking system support traceability, automation, and SBS-format compatibility.
  • 4.0 mL capacity.
  • Three-code marking system with digital code, 2D barcode on bottom, and 1D barcode on side.
  • External threaded vial body with caps separated (bulk packaging).
  • Packaged 48 per pack, 480 per case.
  • Medical-grade cryogenic polypropylene (PP) meeting USP Class VI standards.
  • Recommended for use in liquid nitrogen (-196 °C).
  • Electron beam sterilized with SAL = 10^-6; DNase/RNase free and non-endotoxin.
  • Meets SBS standard and compatible with automated Brooks system workflows.
Catalog No. 50-273-3349