Learn More
STEMCELL Technologies Cryogenic Vials with Internal Thread, 1 mL, Size: 100 Units
SDP
Supplier:  STEMCELL Technologies 2000660
Encompass
Achieve reliable sample cryostorage with Cryogenic Vials with Internal Thread. These sterilized clear polypropylene vials withstand temperatures as low as -196C (in gas/vapor phase) and have a self-standing design. With graduated markings and frosted side panels for easy labeling these vials have a polypropylene cap featuring a silicone O-ring for a dependable seal.
Catalog No. 50-239-0391