Learn More
Electron Microscopy Sciences Cryogenic Grid Box Round with Lid, White
SDP
Supplier:  Electron Microscopy Sciences 7116610W
Encompass_Preferred
These specimen grid boxes are tools for storing or transferring cryogenic TEM specimen grids. Features four diamond shaped slots. Non-static clear covershield held in place with stainless steel screw, which is tapped in the center of the box. Box fits the FEI Vitrobot™, Gatan 626™, Gatan 3500™.
Color: White
Catalog No. NC1118100