Learn More
Electron Microscopy Sciences CryoJane Tape-Thick Transfer System, 110 V
SDP
Supplier:  Electron Microscopy Sciences 6280010
Encompass_Preferred
The CryoJane Tape-Transfer System creates frozen sections as thin as 2 microns with the same quality as sections cut from paraffin blocks. Sections are wrinkle-free, uncompressed, and fully intact when bonded to the microscope slide. The CryoJane uses adhesive coated slides and adhesive tapes to capture sections instead of using an anti-roll plate or brush. CryoJane is suitable for routine cryosections and is indispensable for sectioning difficult tissues including fatty breast and undecalcified bone.
Thin tissue sections remain unflawed even after mounting and this makes CryoJane ideal for producing perfectly thin sections to clarify structural relationships and reveal morphological detail. The CryoJane Tape-Transfer System is adaptable to most cryostats and can be used for histological stains, immuno stains, in situ hybridization, laser microdissection and laser capture microdissection, and frozen tissue arrays.
Catalog No. 50-304-13