Learn More
Cell Applications Inc Human Aortic Smooth Muscle Cells Total Kit
SDP
Supplier:  Cell Applications Inc 354K05A
Encompass
Human Aortic Smooth Muscle Cells Total Kit:(354-05a) Cryopreserved HAOSMC 1 vial, adult(311-500) Smooth Muscle Cell Growth Media 500ml(090K) Subculture Reagent Kit
Catalog No. 50-195-9089