Learn More
Sarstedt Inc CRYOPURE,2ML,EXT RED CAP/PK500
Supplier:  Sarstedt Inc 72.379.002
Encompass_Preferred
Tube, Cyrogenic; CyroPure; Sarstedt; Vol.: 1.8mL; With external thread; Sterile; Red cap; Interlocking base enables single-handed operation in CryoRack 40 and most cryogenic racks; Fit standard cryo boxes and aluminium holders; 500/pk.
Catalog No. 50-873-795