Learn More
Key Scientific Products CRYOSAFE 12 Place Freezer Rack
SDP
Supplier:  Key Scientific Products KS18847
Encompass_Preferred
The CRYOSAFE 12 place freezer rack saves freezer space and prevents damage to enzymes or cultures in frost-free freezers. It will maintain a -15C temperature for 1.5 hours and will remain below 0C for four hours when frozen in a freezer set at -20C. CryoSafe holds 12 tubes with an I.D. up to 12.4mm, such as CryoCare or other brands.
Catalog No. NC9475604