Learn More
Medchemexpress LLC Cryosim-3 | 1507344-37-7 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1265931MG
Encompass_Preferred
Cryosim-3 is a selective TRPM8 receptor agonist that has been shown to increase tear secretion and relieve ocular discomfort in animal models.
  • Selective TRPM8 receptor agonist
  • Increases tear secretion
  • Relieves ocular discomfort in animal models
Catalog No. 50-003-40101