Learn More
Medchemexpress LLC Cryosim-3 | 1507344-37-7 | 99.9% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12659310MG
Encompass_Preferred
Cryosim-3 is a selective TRPM8 receptor agonist. It increases tear secretion and relieves ocular discomfort in animal models. This product is for research use only and not for sale to patients.
  • Selective TRPM8 receptor agonist.
  • Increases tear secretion.
  • Relieves ocular discomfort in animal models.
  • High purity of 99.9%.
  • Stable under recommended storage conditions.
  • Suitable for various research applications.
Catalog No. 50-003-42603