Learn More
Mitegen CryoSleeve for Cryo Storage Canes, Polyvinyl Chloride, Enhanced Security during Handling & Storage, 12 Sleeves per Pack
SDP
Supplier:  Mitegen MCC305
Encompass
The CryoSleeve is a transparent protective sleeve meticulously designed for enclosing Cryo Storage Canes (SKU M-CC-301) , providing enhanced security during both handling and long-term storage of cryogenic samples. Crafted from durable, clear polyvinyl chloride, this sleeve adds an extra layer of protection, safeguarding valuable samples and preventing accidental dislodgement from canes in liquid nitrogen (LN2) dewars or bulk storage tanks. Its transparent material allows for easy visual inspection of the cane's contents without removal. This sleeve is an ideal solution for laboratories seeking to optimize the security and organization of their cryo-sample inventory. By providing an additional containment layer, the CryoSleeve helps minimize risks of contamination and physical damage, ensuring sample integrity. The product is conveniently supplied as a package of 12 sleeves, offering ample supply for various laboratory needs.
Catalog No. NC1769883