Learn More
Akron Biotechnology LLC CryoSolve Syringe Kit, DMSO 55%, GMP Grade, 25 x 7 mL Syringes, AK8217-2507
Catalog No. NC3073805
Encompass
Akron’s CryoSolve product is manufactured following all relevant cGMP guidelines for ancillary materials. The formulation contains sterile, filtered DMSO (Dimethyl Sulfoxide, USP/EP) and Dextran-40 (USP/EP/JP).Product Features: Manufacturing• Manufactured in compliance with cGMP guidelines• In-house sterile filtration and aseptic filling• No animal-derived materials are used in the manufacture of this product• Meets BLA requirements for ancillary materials used in cell therapy productStability• 5-year shelf life• Store at 2-8 °C• Transport with cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.
Catalog No. NC3073805 Supplier Akron Biotechnology LLC Supplier No. AK82172507
Akron’s CryoSolve product is manufactured following all relevant cGMP guidelines for ancillary materials. The formulation contains sterile, filtered DMSO (Dimethyl Sulfoxide, USP/EP) and Dextran-40 (USP/EP/JP).Product Features: Manufacturing• Manufactured in compliance with cGMP guidelines• In-house sterile filtration and aseptic filling• No animal-derived materials are used in the manufacture of this product• Meets BLA requirements for ancillary materials used in cell therapy productStability• 5-year shelf life• Store at 2-8 °C• Transport with cold packsFor Use StatementFor research use or further manufacturing use in ex vivo cell therapy applications. This product is not intended for direct in vivo use or for direct clinical use as a drug, therapeutic, biologic, or medical device.