Learn More
Charles River Labs Cell Solutions CryoStor CS2 Biopreservation Media, 100mL Bottle
Supplier:  Charles River Labs Cell Solutions BPM 202102
Encompass
CryoStor is a uniquely formulated serum-free and animal component-free cryopreservation medium containing dimethyl sulfoxide (DMSO). CryoStor is designed to preserve and protect cells during the freezing and thawing processes as well as during storage at low temperature environments (-80°C to -196°C). CryoStor is cGMP manufactured with USP grade components.
Catalog No. 50-225-6132
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov