Learn More
Origen Biomedical CryoStore 250 EVA Freezing Bag, 30 – 70ml, “Standard” Tube Set, 24/case
Supplier:  Origen Biomedical CS250
Encompass
CryoStore 250 EVA Freezing Bag, 30 – 70ml, “Standard” Tube Set. Two female luers with caps
Catalog No. NC9559236