Learn More
Research Products International Corp Cryo-Tags Labels, Laser Sheet, 1.5 x 0.75 Inches, Orange, 1,200 per Package
SDP
Supplier:  Research Products International Corp 247133O
Encompass
Convenient Cryo-Tags offer superior temperature and chemical resistance. Durable labels withstand cryogenic liquid nitrogen storage, boiling water baths, solvents, caustic solutions and other laboratory chemicals. Cryo-Tags fit bottles, flasks, vials, cryoboxes and more. Chemically inert poly vinyl labels adhere to all polypropylene and polyethylene surfaces without cracking, peeling or degrading and can be marked with a regular pen or marker. Laser Tags are available 60 labels per sheet, 20 sheets per package.
Catalog No. 50-136-7958