Learn More
MOLECULAR DEPOT LLC Cryptdin-4 Mouse - 100 ug
Supplier:  MOLECULAR DEPOT LLC MDP0853
Encompass_Preferred
Cryptdin-4 (Mouse) is a synthetic peptide consisting of the sequence Gly-Leu-Leu-Cys-Tyr-Cys-Arg-Lys-Gly-His-Cys-Lys-Arg-Gly-Glu-Arg-Val-Arg-Gly-Thr-Cys-Gly-Ile-Arg-Phe-Leu-Tyr-Cys-Cys-Pro-Arg-Arg (Disulfide bonds between Cys4-Cys29 Cys6-Cys21 and Cys11-Cys28) The molecular weight of this peptide is 3755 2 g/mol and the CAS number is 100742-61-8 This product is for in vitro research use only This product is not for human or animal use
Catalog No. 50-000-13592