Learn More
Medchemexpress LLC Cryptochlorogenic acid | 905-99-7 | 354.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN07875MG
Encompass_Preferred
Cryptochlorogenic acid is a naturally occurring phenolic acid compound with oral effectiveness, demonstrating anti-inflammatory, antioxidant, and anti-cardiac hypertrophy effects. It works by regulating proinflammatory factor expression, inhibiting NF-κB activity, promoting Nrf2 nuclear transfer, and modulating the PI3K/Akt/mTOR/HIF-1α signaling pathway to alleviate physiological stress responses.
  • Naturally occurring phenolic acid compound.
  • Oral effectiveness.
  • Demonstrates anti-inflammatory effects.
  • Acts as an antioxidant.
  • Exhibits anti-cardiac hypertrophy effects.
  • Regulates proinflammatory factor expression.
  • Inhibits NF-κB activity.
  • Promotes Nrf2 nuclear transfer.
  • Modulates the PI3K/Akt/mTOR/HIF-1α signaling pathway.
  • Shows low toxicity to RAW264.7 cells.
  • Inhibits LPS-induced inflammation.
  • Reduces myocardial hypertrophy in H9c2 cells.
Catalog No. 50-004-17182