Learn More
Abcam Cryptotanshinone, STAT3 inhibitor, 10MG
Supplier:  Abcam AB12066610MG
Encompass_Preferred
MW 296.37 g/mol, Purity >97%. STAT3 inhibitor (IC₅₀ = 4.6 μM). Cell-permeable, naturally occuring constitutent of Salvia miltiorrhiza with diverse biological activities. Shows anticancer, antibacterial, anti-inflammatory, antidiabetes and antiobesity activity. Attenuates amyloid plaque deposition in the brain.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7368