Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291506
Encompass
HR2-915-06/Crystal Screen 2 Cryo #06 (E06), 185 ml - 0.65 M Imidazole pH 7.0, 35% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-3