Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291526
Encompass
HR2-915-26/Crystal Screen 2 Cryo #26 (G02), 185 ml - 0.18 M Ammonium sulfate, 0.09 M MES monohydrate pH 6.5, 27% w/v Polyethylene glycol monomethyl ether 5,000, 10% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-158-3