Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291509
Encompass
HR2-915-09/Crystal Screen 2 Cryo #09 (E09), 185 ml - 0.075 M Sodium acetate trihydrate pH 4.6, 1.5 M Sodium chloride, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-6