Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291543
Encompass
HR2-915-43/Crystal Screen 2 Cryo #43 (H07), 185 ml - 0.2 M Ammonium phosphate monobasic, 0.1 M Tris pH 8.5, 50% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-160-0