Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291507
Encompass
HR2-915-07/Crystal Screen 2 Cryo #07 (E07), 185 ml - 8% w/v Polyethylene glycol 1,000, 8% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-156-4