Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291522
Encompass
HR2-915-22/Crystal Screen 2 Cryo #22 (F10), 185 ml - 0.065 M MES monohydrate pH 6.5, 7.8% w/v Polyethylene glycol 20,000, 35% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-157-9