Learn More
Hampton Research CRYSTAL SCREEN 2 CRYO NO185ML
SDP
Supplier:  Hampton Research HR291540
Encompass
HR2-915-40/Crystal Screen 2 Cryo #40 (H04), 185 ml - 0.075 M Tris pH 8.5, 18.75% v/v tert-Butanol, 25% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-159-7