Learn More
Hampton Research CRYSTAL SCREEN 2 NO29 185ML
SDP
Supplier:  Hampton Research HR291229
Encompass
HR2-912-29/Crystal Screen 2 #29 (G05), 185 ml - 0.5 M Ammonium sulfate, 0.1 M HEPES pH 7.5, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-148-8