Learn More
Hampton Research CRYSTAL SCREEN 2 NO40 185ML
SDP
Supplier:  Hampton Research HR291240
Encompass
HR2-912-40/Crystal Screen 2 #40 (H04), 185 ml - 0.1 M Tris pH 8.5, 25% v/v tert-Butanol. Please note items from this supplier are non-returnable.
Catalog No. 50-149-9