Learn More
Hampton Research CRYSTAL SCREEN CRYO NO10 185ML
SDP
Supplier:  Hampton Research HR291410
Encompass
HR2-914-10/Crystal Screen Cryo #10 (A10), 185 ml - 0.17 M Ammonium acetate, 0.085 M Sodium acetate trihydrate pH 4.6, 25.5% w/v Polyethylene glycol 4,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-151-7