Learn More
Hampton Research CRYSTAL SCREEN CRYO NO15 185ML
SDP
Supplier:  Hampton Research HR291415
Encompass
HR2-914-15/Crystal Screen Cryo #15 (B3), 185 ml - 0.17 M Ammonium sulfate, 0.085 M Sodium cacodylate trihydrate pH 6.5, 25.5% w/v Polyethylene glycol 8,000, 15% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-2