Learn More
Hampton Research CRYSTAL SCREEN CRYO NO18 185ML
SDP
Supplier:  Hampton Research HR291418
Encompass
HR2-914-18/Crystal Screen Cryo #18 (B6), 185 ml - 0.16 M Magnesium acetate tetrahydrate, 0.08 M Sodium cacodylate trihydrate pH 6.5, 16% w/v Polyethylene glycol 8,000, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-5