Learn More
Hampton Research CRYSTAL SCREEN CRYO NO21 185ML
SDP
Supplier:  Hampton Research HR291421
Encompass
HR2-914-21/Crystal Screen Cryo #21 (B9), 185 ml - 0.2 M Magnesium acetate tetrahydrate, 0.1 M Sodium cacodylate trihydrate pH 6.5, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-152-8