Learn More
Hampton Research CRYSTAL SCREEN CRYO NO47 185ML
SDP
Supplier:  Hampton Research HR291447
Encompass
HR2-914-47/Crystal Screen Cryo #47 (D11), 185 ml - 0.08 M Sodium acetate trihydrate pH 4.6, 1.6 M Ammonium sulfate, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-4