Learn More
Hampton Research CRYSTAL SCREEN CRYO NO48 185ML
SDP
Supplier:  Hampton Research HR291448
Encompass
HR2-914-48/Crystal Screen Cryo #48 (D12), 185 ml - 0.08 M TRIS hydrochloride pH 8.5, 1.6 M Ammonium phosphate monobasic, 20% v/v Glycerol. Please note items from this supplier are non-returnable.
Catalog No. 50-155-5