Learn More
Hampton Research CRYSTAL SCREEN LITE NO13 185ML
SDP
Supplier:  Hampton Research HR291613
Encompass
HR2-916-13/Crystal Screen Lite #13/MembFac HT (F01), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M TRIS hydrochloride pH 8.5, 15% v/v Polyethylene glycol 400. Please note items from this supplier are non-returnable.
Catalog No. 50-161-8