Learn More
Hampton Research CRYSTAL SCREEN LITE NO25 185ML
SDP
Supplier:  Hampton Research HR291625
Encompass
HR2-916-25/Crystal Screen Lite #25/MembFac HT (G01), 185 ml - 0.1 M Imidazole pH 6.5, 0.5 M Sodium acetate trihydrate. Please note items from this supplier are non-returnable.
Catalog No. 50-163-0