Learn More
Hampton Research CRYSTAL SCREEN LITE NO27 185ML
SDP
Supplier:  Hampton Research HR291627
Encompass
HR2-916-27/Crystal Screen Lite #27/MembFac HT (G03), 185 ml - 0.2 M Sodium citrate tribasic dihydrate, 0.1 M HEPES sodium pH 7.5, 10% v/v 2-Propanol. Please note items from this supplier are non-returnable.
Catalog No. 50-163-2