Learn More
Hampton Research CRYSTAL SCREEN LITE NO34 185ML
SDP
Supplier:  Hampton Research HR291634
Encompass
HR2-916-34/Crystal Screen Lite #34/MembFac HT (G10), 185 ml - 0.1 M Sodium acetate trihydrate pH 4.6, 1.0 M Sodium formate. Please note items from this supplier are non-returnable.
Catalog No. 50-163-9