Learn More
Hampton Research CRYSTAL SCREEN LITE NO40 185ML
SDP
Supplier:  Hampton Research HR291640
Encompass
HR2-916-40/Crystal Screen Lite #40/MembFac HT (H04), 185 ml - 0.1 M Sodium citrate tribasic dihydrate pH 5.6, 10% v/v 2-Propanol, 10% w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-164-5