Learn More
Hampton Research CRYSTAL SCREEN NO01 185ML
SDP
Supplier:  Hampton Research HR291001
Encompass
HR2-910-01/Crystal Screen #01 (A1), 185 ml - 0.02 M Calcium chloride dihydrate, 0.1 M Sodium acetate trihydrate pH 4.6, 30% v/v (+/-)-2-Methyl-2,4-pentanediol. Please note items from this supplier are non-returnable.
Catalog No. 50-141-0